Imprimer

Anti-ARF6 Mouse monoclonal antibody [clone: SPM371]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB19871-100EA 572 EUR
ABCAAB19871-100
Anti-ARF6 Mouse monoclonal antibody [clone: SPM371]
Type: Primary
Antigen: ARF6 (ADP-ribosylation factor 6)
Clonality: monoclonal
Clone: SPM371
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR