Imprimer

Anti-Clostridium difficile Toxin A Mouse monoclonal antibody [clone: PCG4]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB19953-100EA 562 EUR
ABCAAB19953-100
Anti-Clostridium difficile Toxin A Mouse monoclonal antibody [clone: PCG4]
Type: Primary
Antigen: Clostridium difficile Toxin A
Clonality: monoclonal
Clone: PCG4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR