Imprimer

Anti-Procainamide Mouse monoclonal antibody [clone: 704]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB19965-100EA 550 EUR
ABCAAB19965-100
Anti-Procainamide Mouse monoclonal antibody [clone: 704]
Type: Primary
Antigen: Procainamide
Clonality: monoclonal
Clone: 704
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR