Imprimer

Anti-ITGA3 Mouse Monoclonal Antibody [clone: F35 177-1]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB20140-50EA 703 EUR
ABCAAB20140-50
Anti-ITGA3 Mouse Monoclonal Antibody [clone: F35 177-1]
Anticorps
Type: Primary
Antigen: ITGA3 (integrin, alpha 3 (antigen CD49C, alpha 3 subunit of VLA-3 receptor))
Clonality: monoclonal
Clone: F35 177-1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR