Imprimer

Anti-ITGA6 Mouse Monoclonal Antibody [clone: MP 4F10]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB20142-100EA 729 EUR
ABCAAB20142-100
Anti-ITGA6 Mouse Monoclonal Antibody [clone: MP 4F10]
Anticorps
Type: Primary
Antigen: ITGA6 (integrin, alpha 6)
Clonality: monoclonal
Clone: MP 4F10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR