Imprimer

Anti-CDK1 Mouse monoclonal antibody [clone: 3E1.1]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB20151-50EA 572 EUR
ABCAAB20151-50
Anti-CDK1 Mouse monoclonal antibody [clone: 3E1.1]
Type: Primary
Antigen: CDK1 (cyclin-dependent kinase 1)
Clonality: monoclonal
Clone: 3E1.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Xenopus laevis
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR