Imprimer

Anti-ERBB3 Mouse Monoclonal Antibody [clone: SGP1]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB20162-50EA 704 EUR
ABCAAB20162-50
Anti-ERBB3 Mouse Monoclonal Antibody [clone: SGP1]
Anticorps
Type: Primary
Antigen: ERBB3 (v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 3)
Clonality: monoclonal
Clone: SGP1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR