Imprimer

Anti-Interferon alpha Mouse Monoclonal Antibody [clone: ST29]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB20200-100EA 708 EUR
ABCAAB20200-100
Anti-Interferon alpha Mouse Monoclonal Antibody [clone: ST29]
Anticorps
Type: Primary
Antigen: Interferon alpha
Clonality: monoclonal
Clone: ST29
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR