Imprimer

Anti-Macrophage Mouse Monoclonal Antibody [clone: 5/9]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB20214-50EA 691 EUR
ABCAAB20214-50
Anti-Macrophage Mouse Monoclonal Antibody [clone: 5/9]
Anticorps
Type: Primary
Antigen: Macrophage
Clonality: monoclonal
Clone: 42133
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR