Imprimer

Anti-SV40 T-antigen Mouse monoclonal antibody [clone: Pab 416]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB20248-100EA 606 EUR
ABCAAB20248-100
Anti-SV40 T-antigen Mouse monoclonal antibody [clone: Pab 416]
Type: Primary
Antigen: SV40 T-antigen
Clonality: monoclonal
Clone: Pab 416
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR