Imprimer

Anti-XRCC3 Mouse Monoclonal Antibody [clone: XRCC3 10F1/6]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB20254-50EA 907 EUR
ABCAAB20254-50
Anti-XRCC3 Mouse Monoclonal Antibody [clone: XRCC3 10F1/6]
Anticorps
Type: Primary
Antigen: XRCC3 (X-ray repair complementing defective repair in Chinese hamster cells 3)
Clonality: monoclonal
Clone: XRCC3 10F1/6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR