Imprimer

Anti-Respiratory Syncytial Virus Goat Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB20531-500EA 698 EUR
ABCAAB20531-500
Anti-Respiratory Syncytial Virus Goat Polyclonal Antibody
Anticorps
Type: Primary
Antigen: Respiratory Syncytial Virus
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR