Imprimer

Anti-Asparaginase Rabbit polyclonal antibody Biotin

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB20646-1EA 550 EUR
ABCAAB20646-1
Anti-Asparaginase Rabbit polyclonal antibody Biotin
Type: Primary
Antigen: Asparaginase
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: E. coli
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR