Imprimer

Anti-Rotavirus p42 Mouse monoclonal antibody [clone: 3C10]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB20736-500EA 565 EUR
ABCAAB20736-500
Anti-Rotavirus p42 Mouse monoclonal antibody [clone: 3C10]
Type: Primary
Antigen: Rotavirus p42
Clonality: monoclonal
Clone: 3C10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR