Imprimer

Anti-Hepatitis B Virus Core Antigen Mouse monoclonal antibody [clone: 10C6]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB20749-100EA 572 EUR
ABCAAB20749-100
Anti-Hepatitis B Virus Core Antigen Mouse monoclonal antibody [clone: 10C6]
Type: Primary
Antigen: Hepatitis B Virus Core Antigen
Clonality: monoclonal
Clone: 10C6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR