Imprimer

Anti-Streptococcus group A Mouse monoclonal antibody [clone: 501]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB20915-100EA 550 EUR
ABCAAB20915-100
Anti-Streptococcus group A Mouse monoclonal antibody [clone: 501]
Type: Primary
Antigen: Streptococcus group A
Clonality: monoclonal
Clone: 501
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR