Imprimer

Anti-Neisseria gonorrhoeae Mouse monoclonal antibody [clone: 804]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB20939-100EA 565 EUR
ABCAAB20939-100
Anti-Neisseria gonorrhoeae Mouse monoclonal antibody [clone: 804]
Type: Primary
Antigen: Neisseria gonorrhoeae
Clonality: monoclonal
Clone: 804
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR