Imprimer

Anti-Sulfadimethoxine Mouse monoclonal antibody [clone: CH-2027]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB20966-100EA 565 EUR
ABCAAB20966-100
Anti-Sulfadimethoxine Mouse monoclonal antibody [clone: CH-2027]
Type: Primary
Antigen: Sulfadimethoxine
Clonality: monoclonal
Clone: CH-2027
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR