Imprimer

Anti-Luteinizing Hormone Mouse monoclonal antibody [clone: B20]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB20977-250EA 572 EUR
ABCAAB20977-250
Anti-Luteinizing Hormone Mouse monoclonal antibody [clone: B20]
Type: Primary
Antigen: Luteinizing Hormone
Clonality: monoclonal
Clone: B20
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR