Imprimer

Anti-APOC3 Goat Polyclonal Antibody (Biotin)

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB21024-100EA 714 EUR
ABCAAB21024-100
Anti-APOC3 Goat Polyclonal Antibody (Biotin)
Anticorps
Type: Primary
Antigen: APOC3 (apolipoprotein C-III)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR