Imprimer

Anti-HSV1 + HSV2 Rabbit polyclonal antibody FITC (Fluorescein)

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB21038-100EA 540 EUR
ABCAAB21038-100
Anti-HSV1 + HSV2 Rabbit polyclonal antibody FITC (Fluorescein)
Type: Primary
Antigen: HSV1 + HSV2
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR