Imprimer

Anti-Valproic Acid Mouse monoclonal antibody [clone: 803]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB21101-500EA 550 EUR
ABCAAB21101-500
Anti-Valproic Acid Mouse monoclonal antibody [clone: 803]
Type: Primary
Antigen: Valproic Acid
Clonality: monoclonal
Clone: 803
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR