Imprimer

Anti-B7 DC Rat Monoclonal Antibody [clone: TY25]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB21107-100EA 707 EUR
ABCAAB21107-100
Anti-B7 DC Rat Monoclonal Antibody [clone: TY25]
Anticorps
Type: Primary
Antigen: B7 DC
Clonality: monoclonal
Clone: TY25
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR