Imprimer

Anti-HHV6 Mouse monoclonal antibody [clone: 300]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB21120-100EA 565 EUR
ABCAAB21120-100
Anti-HHV6 Mouse monoclonal antibody [clone: 300]
Type: Primary
Antigen: HHV6
Clonality: monoclonal
Clone: 300
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR