Imprimer

Anti-Hepatitis A Virus Mouse monoclonal antibody [clone: 581]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB21140-100EA 575 EUR
ABCAAB21140-100
Anti-Hepatitis A Virus Mouse monoclonal antibody [clone: 581]
Type: Primary
Antigen: Hepatitis A Virus
Clonality: monoclonal
Clone: 581
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR