Imprimer

Anti-Chlamydia Pneumoniae Mouse monoclonal antibody [clone: 165]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB21175-100EA 540 EUR
ABCAAB21175-100
Anti-Chlamydia Pneumoniae Mouse monoclonal antibody [clone: 165]
Type: Primary
Antigen: Chlamydia Pneumoniae
Clonality: monoclonal
Clone: 165
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR