Imprimer

Anti-LPL Mouse Monoclonal Antibody [clone: LPL.A4]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB21356-100EA 735 EUR
ABCAAB21356-100
Anti-LPL Mouse Monoclonal Antibody [clone: LPL.A4]
Anticorps
Type: Primary
Antigen: LPL (lipoprotein lipase)
Clonality: monoclonal
Clone: LPL.A4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR