Imprimer

Anti-CCL2 Armenian Hamster monoclonal antibody unconjugated [clone: 2H5]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB21396-100EA 724 EUR
ABCAAB21396-100
Anti-CCL2 Armenian Hamster monoclonal antibody unconjugated [clone: 2H5]
Anticorps
Type: Primary
Antigen: CCL2 (chemokine (C-C motif) ligand 2)
Clonality: monoclonal
Clone: 2H5
Conjugation: unconjugated
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR