Imprimer

Anti-ELN Mouse monoclonal antibody [clone: T11E3]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB21598-200EA 586 EUR
ABCAAB21598-200
Anti-ELN Mouse monoclonal antibody [clone: T11E3]
Type: Primary
Antigen: ELN (elastin)
Clonality: monoclonal
Clone: T11E3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR