Imprimer

Anti-WDFY1 Goat polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB21695-50EA 540 EUR
ABCAAB21695-50
Anti-WDFY1 Goat polyclonal antibody
Type: Primary
Antigen: WDFY1 (WD repeat and FYVE domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR