Imprimer

Anti-TUBB4A Mouse polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB21754-100EA 550 EUR
ABCAAB21754-100
Anti-TUBB4A Mouse polyclonal antibody
Type: Primary
Antigen: TUBB4A (tubulin, beta 4A class IVa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR