Imprimer

Anti-ITGB1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: MEM-101A]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB21845-50EA 455 EUR
ABCAAB21845-50
Anti-ITGB1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: MEM-101A]
Anticorps
Type: Primary
Antigen: ITGB1 (integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12))
Clonality: monoclonal
Clone: MEM-101A
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR