Imprimer

Anti-ITGA2B Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: MEM-06]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB21851-50EA 877 EUR
ABCAAB21851-50
Anti-ITGA2B Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: MEM-06]
Anticorps
Type: Primary
Antigen: ITGA2B (integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41))
Clonality: monoclonal
Clone: MEM-06
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR