Imprimer

Anti-PIWIL4 Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB21869-100EA 877 EUR
ABCAAB21869-100
Anti-PIWIL4 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: PIWIL4 (piwi-like RNA-mediated gene silencing 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR