Imprimer

Anti-CD34 Mouse Monoclonal Antibody (Biotin) [clone: 4H11[APG]]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB21893-100EA 698 EUR
ABCAAB21893-100
Anti-CD34 Mouse Monoclonal Antibody (Biotin) [clone: 4H11[APG]]
Anticorps
Type: Primary
Antigen: CD34 (CD34 molecule)
Clonality: monoclonal
Clone: 4H11[APG]
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR