Imprimer

Anti-BSG Mouse monoclonal antibody [clone: MEM-M6/2]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB21903-100EA 632 EUR
ABCAAB21903-100
Anti-BSG Mouse monoclonal antibody [clone: MEM-M6/2]
Type: Primary
Antigen: BSG (basigin (Ok blood group))
Clonality: monoclonal
Clone: MEM-M6/2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Pig
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR