Imprimer

Anti-INA Mouse monoclonal antibody [clone: 2E4]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB22040-100EA 540 EUR
ABCAAB22040-100
Anti-INA Mouse monoclonal antibody [clone: 2E4]
Type: Primary
Antigen: INA (internexin neuronal intermediate filament protein, alpha)
Clonality: monoclonal
Clone: 20000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR