Imprimer

Anti-TLR4 Mouse Monoclonal Antibody [clone: 76B357.1]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB22048-100EA 749 EUR
ABCAAB22048-100
Anti-TLR4 Mouse Monoclonal Antibody [clone: 76B357.1]
Anticorps
Type: Primary
Antigen: TLR4 (toll-like receptor 4)
Clonality: monoclonal
Clone: 76B357.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR