Imprimer

Anti-TLR11 Mouse monoclonal antibody [clone: 15F1215]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB22051-100EA 550 EUR
ABCAAB22051-100
Anti-TLR11 Mouse monoclonal antibody [clone: 15F1215]
Type: Primary
Antigen: TLR11
Clonality: monoclonal
Clone: 15F1215
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR