Imprimer

Anti-SARS Nucleocapsid Protein Mouse monoclonal antibody [clone: 18F629.1]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB22052-100EA 550 EUR
ABCAAB22052-100
Anti-SARS Nucleocapsid Protein Mouse monoclonal antibody [clone: 18F629.1]
Type: Primary
Antigen: SARS Nucleocapsid Protein
Clonality: monoclonal
Clone: 18F629.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR