Imprimer

Anti-TNFSF11 Rabbit polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB22113-100EA 609 EUR
ABCAAB22113-100
Anti-TNFSF11 Rabbit polyclonal antibody
Type: Primary
Antigen: TNFSF11 (tumor necrosis factor (ligand) superfamily, member 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR