Imprimer

Anti-YAP1 Mouse polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB22144-100EA 606 EUR
ABCAAB22144-100
Anti-YAP1 Mouse polyclonal antibody
Type: Primary
Antigen: YAP1 (Yes-associated protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR