Imprimer

Anti-HPS4 Mouse polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB22147-100EA 565 EUR
ABCAAB22147-100
Anti-HPS4 Mouse polyclonal antibody
Type: Primary
Antigen: HPS4 (Hermansky-Pudlak syndrome 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR