Imprimer

Anti-ALAS1 Rabbit polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB22153-100EA 579 EUR
ABCAAB22153-100
Anti-ALAS1 Rabbit polyclonal antibody
Type: Primary
Antigen: ALAS1 (aminolevulinate, delta-, synthase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR