Imprimer

Anti-MAPT Mouse monoclonal antibody [clone: Tau 46]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB22261-100EA 572 EUR
ABCAAB22261-100
Anti-MAPT Mouse monoclonal antibody [clone: Tau 46]
Type: Primary
Antigen: MAPT (microtubule-associated protein tau)
Clonality: monoclonal
Clone: Tau 46
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR