Imprimer

Anti-REL Rabbit polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB22263-100EA 540 EUR
ABCAAB22263-100
Anti-REL Rabbit polyclonal antibody
Type: Primary
Antigen: REL (v-rel avian reticuloendotheliosis viral oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR