Imprimer

Anti-ICAM1 Mouse monoclonal antibody [clone: 1A29]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB22388-100EA 550 EUR
ABCAAB22388-100
Anti-ICAM1 Mouse monoclonal antibody [clone: 1A29]
Type: Primary
Antigen: ICAM1 (intercellular adhesion molecule 1)
Clonality: monoclonal
Clone: 1A29
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR