Imprimer

Anti-Poliovirus (type 1-3) Goat Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB22450-500EA 878 EUR
ABCAAB22450-500
Anti-Poliovirus (type 1-3) Goat Polyclonal Antibody
Anticorps
Type: Primary
Antigen: Poliovirus (type 1-3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR