Imprimer

Anti-HSV1 gG Envelope Protein Mouse monoclonal antibody [clone: BO18]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB22481-100EA 550 EUR
ABCAAB22481-100
Anti-HSV1 gG Envelope Protein Mouse monoclonal antibody [clone: BO18]
Type: Primary
Antigen: HSV1 gG Envelope Protein
Clonality: monoclonal
Clone: BO18
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR