Imprimer

Anti-CD4 Rat monoclonal antibody [clone: KT174]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB22496-1EA 540 EUR
ABCAAB22496-1
Anti-CD4 Rat monoclonal antibody [clone: KT174]
Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: monoclonal
Clone: KT174
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2c
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR